missing translation for 'onlineSavingsMsg'
Få mere at vide

3-Ethylbenzaldehyde, TRC

Artikelnummer. 30380303
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30380303 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30380303 Leverandør Toronto Research Chemicals Leverandørnr. TRCE899838500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 3-Ethylbenzaldehyde
CAS 34246-54-3
Molekylær formel C9H10O
Mængde 500 mg
InChI formel InChI=1S/C9H10O/c1-2-8-4-3-5-9(6-8)7-10/h3-7H,2H2,1H3
SMIL CCC1=CC(=CC=C1)C=O
IUPAC navn 3-ethylbenzaldehyde
Molekylvægt (g/mol) 134.17
Formel vægt 134.0732
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.