Få mere at vide
3-Ethyl Adenine-d5 (2% d0), TRC
Artikelnummer. 30378347
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30378347 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378347 Leverandør Toronto Research Chemicals Leverandørnr. TRCE8971501MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Ethyl-d5-adenine
CAS 147028-85-1
Molekylær formel C7 2H5 H4 N5
Mængde 1 mg
Synonym 3-Ethyl Adenine-D5
InChI formel InChI=1S/C7H9N5/c1-2-12-4-11-6(8)5-7(12)10-3-9-5/h3-4H,2,8H2,1H3/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])n1cnc(N)c2ncnc12
IUPAC navn 3-(1,1,2,2,2-pentadeuterioethyl)purin-6-amine
Molekylvægt (g/mol) 168.21
Formel vægt 168.1172