Learn More
3-Ethyl-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
Catalog Number p-7030043
Click to view available options
Quantity:
1 g
10 g
17147-85-2
C7H9NO3
155.153
MFCD03422293
RSWFHHWVFZWIMV-UHFFFAOYSA-N
3-ethyl-5-methylisoxazole-4-carboxylic acid, 3-ethyl-5-methyl-isoxazole-4-carboxylic acid, 3-ethyl-5-methyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-ethyl-5-methyl, acmc-1c8pz, 4-isoxazolecarboxylicacid,3-ethyl-5-methyl, 4-isoxazolecarboxylicacid, 3-ethyl-5-methyl, 4-isoxazolecarboxylic acid, 3-ethyl-5-methyl-8ci,9ci
2063310
3-ethyl-5-methyl-1,2-oxazole-4-carboxylic acid
CCC1=NOC(=C1C(=O)O)C
17147-85-2
C7H9NO3
155.153
MFCD03422293
RSWFHHWVFZWIMV-UHFFFAOYSA-N
3-ethyl-5-methylisoxazole-4-carboxylic acid, 3-ethyl-5-methyl-isoxazole-4-carboxylic acid, 3-ethyl-5-methyl-4-isoxazolecarboxylic acid, 4-isoxazolecarboxylic acid, 3-ethyl-5-methyl, acmc-1c8pz, 4-isoxazolecarboxylicacid,3-ethyl-5-methyl, 4-isoxazolecarboxylicacid, 3-ethyl-5-methyl, 4-isoxazolecarboxylic acid, 3-ethyl-5-methyl-8ci,9ci
2063310
3-ethyl-5-methyl-1,2-oxazole-4-carboxylic acid
CCC1=NOC(=C1C(=O)O)C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 123°C to 125°C
Quantity 1 g
Formula Weight 155.15
Percent Purity 97%
Chemical Name or Material 3-Ethyl-5-methylisoxazole-4-carboxylic acid