missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethoxythiophene-2-carboxylic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3-Ethoxythiophene-2-carboxylic acid is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Ethoxythiophene-2-carboxylic acid is used as a pharmaceutical intermediate.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, bases, reducing agents.
3-Ethoxythiophene-2-carboxylic acid is used as a pharmaceutical intermediate.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, bases, reducing agents.
Specifications
Melting Point | 121°C to 123°C |
Quantity | 1 g |
Synonym | maybridge1_003798, acmc-20an4i, 3-ethoxy-2-thiophenecarboxylic acid, 2-thiophenecarboxylicacid,3-ethoxy, 2-thiophenecarboxylicacid, 3-ethoxy, 2-thiophenecarboxylic acid, 3-ethoxy |
Formula Weight | 172.2 |
Percent Purity | 97% |
Chemical Name or Material | 3-Ethoxythiophene-2-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'