missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-5-fluorobenzeneboronic acid, 95%, Thermo Scientific™

Catalog Number p-7034752
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
850589-53-6
C8H10BFO3
183.97
MFCD07363779
OVVBLFYHZAMJIK-UHFFFAOYSA-N
3-ethoxy-5-fluorobenzeneboronic acid, 3-ethoxy-5-fluorophenyl boronic acid, 5-ethoxy-3-fluorophenylboronic acid, boronic acid,b-3-ethoxy-5-fluorophenyl, 3-ethoxy-5-fluorophenylboronicacid, acmc-209q3b, 3-ethoxy-5-fluoro-benzeneboronic acid, 3-ethoxy-5-fluoro-phenyl boronic acid, 3-ethoxy-5-fluorobenzeneboronicacid, 3-ethoxy-5-fluoranyl-phenyl boronic acid
44558175
(3-ethoxy-5-fluorophenyl)boronic acid
CCOC1=CC(F)=CC(=C1)B(O)O
This item is not returnable. View return policy
850589-53-6
C8H10BFO3
183.97
MFCD07363779
OVVBLFYHZAMJIK-UHFFFAOYSA-N
3-ethoxy-5-fluorobenzeneboronic acid, 3-ethoxy-5-fluorophenyl boronic acid, 5-ethoxy-3-fluorophenylboronic acid, boronic acid,b-3-ethoxy-5-fluorophenyl, 3-ethoxy-5-fluorophenylboronicacid, acmc-209q3b, 3-ethoxy-5-fluoro-benzeneboronic acid, 3-ethoxy-5-fluoro-phenyl boronic acid, 3-ethoxy-5-fluorobenzeneboronicacid, 3-ethoxy-5-fluoranyl-phenyl boronic acid
44558175
(3-ethoxy-5-fluorophenyl)boronic acid
CCOC1=CC(F)=CC(=C1)B(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 155°C to 158°C
Quantity 250 mg
Formula Weight 183.97
Percent Purity 95%
Chemical Name or Material 3-Ethoxy-5-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'