Få mere at vide
3-epi-Pregnenolone, TRC
Artikelnummer. 30418817
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418817 2.5 mg
30413748 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30418817 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7122052.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 19037-28-6
Molekylær formel C21 H32 O2
Molekylvægt (g/mol) 316.48
Synonym Pregn-5-en-20-one, 3-hydroxy-, (3α)-, Pregn-5-en-20-one, 3α-hydroxy- (8CI), (3α)-3-Hydroxypregn-5-en-20-one, 3α-Hydroxy-20-oxopregn-5-ene, Pregn-5-en-3α-ol-20-one, δ5-Pregnen-3α-ol-20-one, 3-epi-Pregnenolone
IUPAC navn 1-[(3R,8S,9S,10R,13S,14S,17S)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
SMIL CC(=O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale 3-epi-Pregnenolone
Mængde 2.5 mg
InChI formel InChI=1S/C21H32O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h4,15-19,23H,5-12H2,1-3H3/t15-,16+,17-,18+,19+,20+,21-/m1/s1
Formel vægt 316.2402