Få mere at vide
3-epi-25-Hydroxy Vitamin D3, TRC
Artikelnummer. 30407093
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30407093 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407093 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9958602.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-epi-25-Hydroxyvitamin D3
CAS 73809-05-9
Renhedsgrad noter HPLC
Molekylær formel C27 H44 O2
Mængde 2.5 mg
Synonym 1H-Indene-1-pentanol, octahydro-4-[(2Z)-2-[(5R)-5-hydroxy-2-methylenecyclohexylidene]ethylidene]-α,α,ε,7a-tetramethyl-, (εR,1R,3aS,4E,7aR)-, 9,10-Secocholesta-5,7,10(19)-triene-3,25-diol, (3α,5Z,7E)- (9CI), (εR,1R,3aS,4E,7aR)-Octahydro-4-[(2Z)-2-[(5R)-5-hydroxy-2-methylenecyclohexylidene]ethylidene]-α,α,ε,7a-tetramethyl-1H-indene-1-pentanol, 3-epi-Calcifediol, 3-epi-25-Hydroxycholecalciferol, 3-epi-25-Hydroxyitamin D3, 3-epi-25-Hydroxyvitamin D3, Vitamin D3-3-epi-25-hydroxy
InChI formel InChI=1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23-,24-,25+,27-/m1/s1
SMIL C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@H](O)CCC3=C)\CCC[C@]12C
IUPAC navn (1R,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(2R)-6-hydroxy-6-methylheptan-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
Molekylvægt (g/mol) 400.64
Formel vægt 400.3341
Procent renhed >95