Få mere at vide
3-Dimethylaminopropyl Chloride Hydrochloride, TRC
Artikelnummer. 30137389
Skift visning
Mængde:
2.5 g
25 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30137389 25 g -
30011597 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137389 Leverandør Toronto Research Chemicals Leverandørnr. TRCD46180025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5407-04-5
Molekylær formel C5 H12 Cl N . Cl H
Molekylvægt (g/mol) 158.07
Synonym N,N-Dimethyl-3-chloropropylamine Hydrochloride, 1-Chloropropyl(dimethyl)amine Hydrochloride, 1-Propanamine, 3-chloro-N,N-dimethyl-, hydrochloride (1:1) (ACI), 1-Propanamine, 3-chloro-N,N-dimethyl-, hydrochloride (9CI), Propylamine, 3-chloro-N,N-dimethyl-, hydrochloride (7CI, 8CI), (3-Chloropropyl)dimethylamine hydrochloride, (3-Chloropropyl)dimethylammonium chloride, 1-Chloro-3-(dimethylamino)propane hydrochloride, 1-Dimethylamino-3-chloropropane hydrochloride, 3-(Dimethylamino)-1-chloropropane hydrochloride, 3-(Dimethylamino)-1-propyl chloride hydrochloride, 3-(Dimethylamino)propyl chloride hydrochloride, 3-Chloro-1-(N,N-dimethylamino)propane hydrochloride, 3-Chloro-N,N-dimethyl-1-propanamine hydrochloride, 3-Chloro-N,N-dimethylpropanamine hydrochloride, 3-Chloro-N,N-dimethylpropane-1-amine hydrochloride, 3-Chloro-N,N-dimethylpropylamine hydrochloride, N,N-Dimethyl-3-chloropropan-1-amine hydrochloride, N,N-Dimethyl-N-(3-chloropropyl)amine hydrochloride, N-(3-Chloropropyl)dimethylamine hydrochloride
IUPAC navn 3-chloro-N,N-dimethylpropan-1-amine;hydrochloride
SMIL Cl.CN(C)CCCCl
Tekniske data
Kemisk navn eller materiale N,N-Dimethyl-3-chloropropylamine Hydrochloride
Mængde 25 g
InChI formel InChI=1S/C5H12ClN.ClH/c1-7(2)5-3-4-6;/h3-5H2,1-2H3;1H
Formel vægt 157.0425