Learn More
3-Dimethylaminophenol, 97%, ACROS Organics™
Brand: Acros Organics 115865000
Additional Details : CAS Number : 99-07-0 Weight : 0.55000kg
Packaging | Glass bottle |
---|---|
Quantity | 500g |
Chemical Identifiers
99-07-0 | |
137.18 | |
MESJRHHDBDCQTH-UHFFFAOYSA-N | |
7421 | |
CN(C)C1=CC(=CC=C1)O |
C8H11NO | |
MFCD00002264 | |
3-dimethylamino phenol, phenol, 3-dimethylamino, m-dimethylamino phenol, m-dimethylaminophenol, n,n-dimethyl-m-aminophenol, n,n-dimethyl-3-aminophenol, phenol, m-dimethylamino, 3-hydroxyphenyl dimethylamine, 3-hydroxy-n,n-dimethylaniline, 3-n,n-dimethylaminophenol | |
3-(dimethylamino)phenol |
Specifications
3-Dimethylaminophenol | |
Authentic | |
Brown to Gray or Purple | |
500g | |
96.0 | |
96% min. (GC) | |
(CH3)2NC6H4OH | |
13, 405 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether | |
CN(C)C1=CC(=CC=C1)O | |
137.18 | |
137.18 | |
97% |
148°C | |
Glass bottle | |
80°C to 85°C | |
99-07-0 | |
100.0 | |
C8H11NO | |
MFCD00002264 | |
3-dimethylamino phenol, phenol, 3-dimethylamino, m-dimethylamino phenol, m-dimethylaminophenol, n,n-dimethyl-m-aminophenol, n,n-dimethyl-3-aminophenol, phenol, m-dimethylamino, 3-hydroxyphenyl dimethylamine, 3-hydroxy-n,n-dimethylaniline, 3-n,n-dimethylaminophenol | |
MESJRHHDBDCQTH-UHFFFAOYSA-N | |
3-(dimethylamino)phenol | |
7421 | |
Powder |
Safety and Handling
- 3-Dimethylaminophenol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.