missing translation for 'onlineSavingsMsg'
Learn More

3-Diethylamino-1-propyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog Number p-7045445
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2626-52-0
C8H16N2S
172.29
MFCD00041133
QPBVKSLJBRCKIF-UHFFFAOYSA-N
3-diethylamino propyl isothiocyanate, diethyl 3-isothiocyanatopropyl amine, 3-diethylamino propylisothiocyanate, diethyl-3-isothiocyanato-propyl-amine, acmc-20aniz, n,n-diethyl-3-isothiocyanato-propan-1-amine, 3-diethylaminopropyl isothiocyanate, 3-diethylamino propanisothiocyanate, 3-diethylamino-1-propyl isothiocyanate, 3-n,n-diethylamino propylisothiocyanate
2758240
N,N-diethyl-3-isothiocyanatopropan-1-amine
CCN(CC)CCCN=C=S
This item is not returnable. View return policy
2626-52-0
C8H16N2S
172.29
MFCD00041133
QPBVKSLJBRCKIF-UHFFFAOYSA-N
3-diethylamino propyl isothiocyanate, diethyl 3-isothiocyanatopropyl amine, 3-diethylamino propylisothiocyanate, diethyl-3-isothiocyanato-propyl-amine, acmc-20aniz, n,n-diethyl-3-isothiocyanato-propan-1-amine, 3-diethylaminopropyl isothiocyanate, 3-diethylamino propanisothiocyanate, 3-diethylamino-1-propyl isothiocyanate, 3-n,n-diethylamino propylisothiocyanate
2758240
N,N-diethyl-3-isothiocyanatopropan-1-amine
CCN(CC)CCCN=C=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.95
Boiling Point 94°C to 95°C (3.5 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Refractive Index 1.503
Quantity 1 g
UN Number UN2922
Beilstein 1757446
Sensitivity Moisture sensitive
Formula Weight 172.29
Percent Purity 97%
Chemical Name or Material 3-Diethylamino-1-propyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'