Få mere at vide
3-Desoxyvitamin D3 (~90%), TRC
Artikelnummer. 30354491
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30354491 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30354491 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29699810MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Desoxyvitamin D3
CAS 69662-75-5
Molekylær formel C27 H44
Mængde 10 mg
Synonym (1R,3aS,7aR,E)-7a-Methyl-4-((Z)-2-(2-methylenecyclohexylidene)ethylidene)-1-((R)-6-methylheptan-2-yl)octahydro-1H-indene
InChI formel InChI=1S/C27H44/c1-20(2)10-8-12-22(4)25-17-18-26-24(14-9-19-27(25,26)5)16-15-23-13-7-6-11-21(23)3/h15-16,20,22,25-26H,3,6-14,17-19H2,1-2,4-5H3/b23-15-,24-16+/t22-,25-,26-,27+/m0/s1
SMIL CC(C)CCC[C@H](C)[C@@H]1CC[C@H]2\C(=C\C=C/3\CCCCC3=C)\CCC[C@]12C
Molekylvægt (g/mol) 368.64