Få mere at vide
3-Descyano Febuxostat Ethyl Ester, TRC
Artikelnummer. 30367336
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367336 1 g
30356008 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30367336 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2897101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 144060-97-9
Molekylær formel C17H21NO3S
Molekylvægt (g/mol) 319.42
Synonym 4-Methyl-2-[4-(2-methylpropoxy)phenyl]-5-thiazolecarboxylic Acid Ethyl Ester, Ethyl 2-(4-Isobutoxyphenyl)-4-methylthiazole-5-carboxylate
IUPAC navn ethyl 4-methyl-2-[4-(2-methylpropoxy)phenyl]-1,3-thiazole-5-carboxylate
SMIL CCOC(=O)c1sc(nc1C)c2ccc(OCC(C)C)cc2
Tekniske data
Kemisk navn eller materiale 3-Descyano Febuxostat Ethyl Ester
Mængde 1 g
InChI formel InChI=1S/C17H21NO3S/c1-5-20-17(19)15-12(4)18-16(22-15)13-6-8-14(9-7-13)21-10-11(2)3/h6-9,11H,5,10H2,1-4H3
Formel vægt 319.1242