Få mere at vide
3-Des(2-methoxyethyl) 3-(1-Methylethyl) Ester Nimodipine, TRC
Artikelnummer. 30133888
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133888 250 mg
30011691 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133888 Leverandør Toronto Research Chemicals Leverandørnr. TRCD291310250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 21881-78-7
Molekylær formel C21 H26 N2 O6
Molekylvægt (g/mol) 402.44
Synonym Bis(1-methylethyl) 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Nimodipine Imp. B (EP)
IUPAC navn dipropan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMIL CC(C)OC(=O)C1=C(C)NC(=C(C1c2cccc(c2)[N+](=O)[O-])C(=O)OC(C)C)C
Tekniske data
Kemisk navn eller materiale Bis(1-methylethyl) 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Renhedsgrad noter HPLC
Mængde 250 mg
InChI formel InChI=1S/C21H26N2O6/c1-11(2)28-20(24)17-13(5)22-14(6)18(21(25)29-12(3)4)19(17)15-8-7-9-16(10-15)23(26)27/h7-12,19,22H,1-6H3
Formel vægt 402.1791
Procent renhed >95%