Få mere at vide
3-Deoxyyunaconitine, TRC
Artikelnummer. 30361625
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361625 100 mg
30354718 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30361625 Leverandør Toronto Research Chemicals Leverandørnr. TRCD281820100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 79592-91-9
Molekylær formel C35 H49 N O10
Molekylvægt (g/mol) 643.76
Synonym (1α,6α,14α,16β)-20-Ethyl-1,6,16-trimethoxy-4-(methoxymethyl)aconitane-8,13,14-triol 8-Acetate 14-(4-methoxybenzoate)
SMIL CCN1C[C@]2(COC)CC[C@H](OC)[C@@]34[C@@H]5C[C@]6(O)[C@H](C[C@](OC(=O)C)([C@H]([C@H](OC)[C@H]23)C14)[C@H]5[C@H]6OC(=O)c7ccc(OC)cc7)OC
Tekniske data
Kemisk navn eller materiale 3-Deoxyyunaconitine
Mængde 100 mg
InChI formel InChI=1S/C35H49NO10/c1-8-36-17-32(18-40-3)14-13-23(42-5)35-22-15-33(39)24(43-6)16-34(46-19(2)37,26(29(35)36)27(44-7)28(32)35)25(22)30(33)45-31(38)20-9-11-21(41-4)12-10-20/h9-12,22-30,39H,8,13-18H2,1-7H3/t22-,23+,24+,25-,26-,27+,28-,29?,30-,32+,33+,34-,35+/m1/s1
Formel vægt 643.3356