Få mere at vide
3'-Deoxyadenosine 5’-Triphosphate Triethylamine Salt, TRC
Artikelnummer. 30367759
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367759 10 mg
30362591 1 mg
30351793 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30367759 Leverandør Toronto Research Chemicals Leverandørnr. TRCD23207010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 110972-45-7
Molekylvægt (g/mol) 895.942
Synonym 3'-Deoxyadenosine 5'-Triphosphate Triethylamine Salt, Adenosine 5'-(tetrahydrogen triphosphate), 3'-deoxy-, compd. with N,N-diethylethanamine (1:4) (9CI), Tetrakis(triethylammonium) 3'-Deoxyadenosine 5'-(tetrahydrogen triphosphate), 3'-Deoxy-ATP triethylammonium salt, 3'-Deoxyadenosine 5'-triphosphate trieth
IUPAC navn [[(2S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;N,N-diethylethanamine
SMIL CCN(CC)CC.CCN(CC)CC.CCN(CC)CC.CCN(CC)CC.Nc1ncnc2c1ncn2[C@@H]3O[C@H](COP(=O)(O)OP(=O)(O)OP(=O)(O)O)C[C@H]3O
Tekniske data
Kemisk navn eller materiale 3'-Deoxyadenosine 5'-Triphosphate Tetra-triethylamine
Molekylær formel C10 H16 N5 O12 P3 . 4 C6 H15 N
Mængde 10 mg
InChI formel InChI=1S/C10H16N5O12P3.4C6H15N/c11-8-7-9(13-3-12-8)15(4-14-7)10-6(16)1-5(25-10)2-24-29(20,21)27-30(22,23)26-28(17,18)19;4*1-4-7(5-2)6-3/h3-6,10,16H,1-2H2,(H,20,21)(H,22,23)(H2,11,12,13)(H2,17,18,19);4*4-6H2,1-3H3/t5-,6+,10+;;;;/m0..../s1
Formel vægt 895.483