Få mere at vide
3'-Deoxy-3'-fluorothymidine-d3, TRC
Artikelnummer. 30367445
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30367445 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30367445 Leverandør Toronto Research Chemicals Leverandørnr. TRCD23547710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3'-Deoxy-3'-fluorothymidine-d3
CAS 1346523-18-9
Molekylær formel C10H10D3FN2O4
Mængde 10 mg
Synonym 1-(3'-Deoxy-3'-fluoro-β-D-pentofuranosyl)thymine-d3, 3'-Fluoro-3'- deoxythymidine-d3, 3'-Fluorodeoxythymidine-d3, 3'-Fluorothymidine-d3, Alovudine-d3, CL 184824-d3, FLT-d3, MIV 310-d3, NSC 140025-d3
InChI formel InChI=1S/C10H13FN2O4/c1-5-3-13(10(16)12-9(5)15)8-2-6(11)7(4-14)17-8/h3,6-8,14H,2,4H2,1H3,(H,12,15,16)/t6-,7+,8+/m0/s1/i1D3
SMIL O=C1C(C([2H])([2H])[2H])=CN([C@H]2C[C@H](F)[C@@H](CO)O2)C(N1)=O
Molekylvægt (g/mol) 247.24