Få mere at vide
3-Deoxy-1,2,4,6-tetra-O-acetyl-D-glucopyranose, TRC
Artikelnummer. 30363037
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363037 50 mg
30349929 250 mg
30356909 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30363037 Leverandør Toronto Research Chemicals Leverandørnr. TRCD27877550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5040-09-5
Molekylær formel C14 H20 O9
Molekylvægt (g/mol) 332.3
IUPAC navn [(2R,3S,5R)-3,5,6-triacetyloxyoxan-2-yl]methyl acetate
SMIL CC(=O)OC[C@H]1OC(OC(=O)C)[C@@H](C[C@@H]1OC(=O)C)OC(=O)C
Tekniske data
Kemisk navn eller materiale 3-Deoxy-1,2,4,6-tetra-O-acetyl-D-glucopyranose
Mængde 50 mg
InChI formel InChI=1S/C14H20O9/c1-7(15)19-6-13-11(20-8(2)16)5-12(21-9(3)17)14(23-13)22-10(4)18/h11-14H,5-6H2,1-4H3/t11-,12+,13+,14?/m0/s1
Formel vægt 332.1107