Få mere at vide
3-Dehydro Retinal, TRC
Artikelnummer. 30364156
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364156 10 mg
30365214 5 mg
30349147 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30364156 Leverandør Toronto Research Chemicals Leverandørnr. TRCD23007510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 472-87-7
Molekylær formel C20H26O
Molekylvægt (g/mol) 282.42
Synonym (all-E)-3,7-Dimethyl-9-(2,6,6-trimethyl-1,3-cyclohexadien-1-yl)-2,4,6,8-nonatetraenal, trans-3-Dehydroretinal, Dehydroretinaldehyde, Retinene 2, Vitamin A2 Aldehyde
IUPAC navn (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexa-1,3-dien-1-yl)nona-2,4,6,8-tetraenal
SMIL CC1=C(C(CC=C1)(C)C)/C=C/C(=C/C=C/C(=C/C=O)/C)/C
Tekniske data
Kemisk navn eller materiale 3-Dehydro Retinal
Mængde 10 mg
InChI formel InChI=1S/C20H26O/c1-16(8-6-9-17(2)13-15-21)11-12-19-18(3)10-7-14-20(19,4)5/h6-13,15H,14H2,1-5H3/b9-6+,12-11+,16-8+,17-13+
Formel vægt 282.1984