Få mere at vide
3-Dechloro-3-bromo Trazodone Hydrochloride, TRC
Artikelnummer. 30350286
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30350286 50 mg
1 options
Artikelnummer. 30350286 Leverandør Toronto Research Chemicals Leverandørnr. TRCD22626050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-3-[4-(3-Bromophenyl)piperazin-1-yl]propyl-1,2,4-triazolo[4,3-a]-pyridin-3(2H)-one Hydrochloride
CAS 1263278-80-3
Molekylær formel C19 H22 Br N5 O . Cl H
Mængde 50 mg
Synonym 1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one, 2-[3-[4-(3-bromophenyl)-1-piperazinyl]propyl]-, hydrochloride (1:1), 2-3-[4-(3-Bromophenyl)piperazin-1-yl]propyl-1,2,4-triazolo[4,3-a]-pyridin-3(2H)-one Hydrochloride, Trazodone Hydrochloride Imp. D (BP) as Hydrochloride, 3-Dechloro-3-bromo Trazodone hydrochloride, 3-Dechloro-3-bromotrazodone hydrochloride
InChI formel InChI=1S/C19H22BrN5O.ClH/c20-16-5-3-6-17(15-16)23-13-11-22(12-14-23)8-4-10-25-19(26)24-9-2-1-7-18(24)21-25;/h1-3,5-7,9,15H,4,8,10-14H2;1H
SMIL Cl.Brc1cccc(c1)N2CCN(CCCN3N=C4C=CC=CN4C3=O)CC2
IUPAC navn 2-[3-[4-(3-bromophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;hydrochloride
Molekylvægt (g/mol) 452.78
Formel vægt 451.0775