missing translation for 'onlineSavingsMsg'
Learn More

3-Deazauridine, TRC

Product Code. 30352134
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30352134 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30352134 Supplier Toronto Research Chemicals Supplier No. TRCD203240100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3-Deazauridine
CAS 23205-42-7
Molekylær formel C10 H13 N O6
Mængde 100 mg
Synonym 2(1H)-Pyridinone, 4-hydroxy-1-β-D-ribofuranosyl-, 2(1H)-Pyridone, 4-hydroxy-1-β-D-ribofuranosyl- (8CI), 4-Hydroxy-1-β-D-ribofuranosyl-2(1H)-pyridinone, 1-β-D-Ribofuranosyl-4-hydroxy-2-pyridone, 3-Deazauridine, 3-Deazuridine, 4-Hydroxy-1-(β-D-ribofuranosyl)-2-pyridone, NSC 126849
InChI formel InChI=1S/C10H13NO6/c12-4-6-8(15)9(16)10(17-6)11-2-1-5(13)3-7(11)14/h1-3,6,8-10,12-13,15-16H,4H2/t6-,8-,9-,10-/m1/s1
SMIL OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC(=CC2=O)O
IUPAC navn 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-4-hydroxypyridin-2-one
Molekylvægt (g/mol) 243.21
Formel vægt 243.0743
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.