Få mere at vide
3-(Cyclopropylcarbamoyl)phenylboronic Acid (>90%), TRC
Artikelnummer. 30366021
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366021 50 mg
30367512 25 mg
30354901 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30366021 Leverandør Toronto Research Chemicals Leverandørnr. TRCC99150350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 850567-23-6
Molekylær formel C10H12BNO3
Molekylvægt (g/mol) 205.02
Synonym B-[3-[(Cyclopropylamino)carbonyl]phenyl]boronic Acid, 3-(Cyclopropylaminocarbonyl)benzeneboronic Acid, 3-(Cyclopropylaminocarbonyl)phenylboronic Acid
IUPAC navn [3-(cyclopropylcarbamoyl)phenyl]boronic acid
SMIL OB(O)c1cccc(c1)C(=O)NC2CC2
Tekniske data
Kemisk navn eller materiale 3-(Cyclopropylcarbamoyl)phenylboronic Acid
Mængde 50 mg
InChI formel InChI=1S/C10H12BNO3/c13-10(12-9-4-5-9)7-2-1-3-8(6-7)11(14)15/h1-3,6,9,14-15H,4-5H2,(H,12,13)
Formel vægt 205.091