missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Cyclopentylcarbamoyl-2-fluorobenzeneboronic acid, 97%

Brand:  Alfa Aesar™ H56079.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402466

  • 4993.71 DKK / 1g


Estimated Shipment date
from Supplier 03-08-2018
Add to basket

Specifications

Chemical Identifiers

3-Cyclopentylcarbamoyl-2-fluorobenzeneboronic acid
MFCD26793809

Specifications

3-Cyclopentylcarbamoyl-2-fluorobenzeneboronic acid
Off-white to pale gray
Powder
MFCD26793809
251.06
1g
97%
Videos
SDS