Få mere at vide
3-cyclobutyl-2,2-dimethylpropanal, TRC
Artikelnummer. 30361109
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361109 500 mg
30351714 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30361109 Leverandør Toronto Research Chemicals Leverandørnr. TRCC230081500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-cyclobutyl-2,2-dimethylpropanal
CAS 1936119-86-6
Molekylær formel C9 H16 O
Mængde 500 mg
InChI formel InChI=1S/C9H16O/c1-9(2,7-10)6-8-4-3-5-8/h7-8H,3-6H2,1-2H3
SMIL CC(C)(CC1CCC1)C=O
IUPAC navn 3-cyclobutyl-2,2-dimethylpropanal
Molekylvægt (g/mol) 140.223
Formel vægt 140.12