Learn More
3-(Cyanomethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-(Cyanomethyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
3-(Cyanomethyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
Melting Point | 44°C to 48°C |
Quantity | 250 mg |
Solubility Information | Slightly soluble in water. |
Formula Weight | 243.11 |
Percent Purity | 96% |
Chemical Name or Material | 3-(Cyanomethyl)benzeneboronic acid pinacol ester |