missing translation for 'onlineSavingsMsg'
Learn More

3-(Cyanomethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15419955
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
396131-82-1
C14H18BNO2
243.11
MFCD02179458
LIWPKYSGWSFPEE-UHFFFAOYSA-N
2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-cyanomethyl phenylboronic acid pinacol ester, 2-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, pubchem7790, 3-cyanomethylphenylboronic acid, pinacol ester, 3-cyanomethylphenyl boronic acid,pinacol ester, 3-cyanomethyl phenylboronic acid, pinacol ester
2773343
2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
CC1(C)OB(OC1(C)C)C1=CC(CC#N)=CC=C1
This item is not returnable. View return policy
396131-82-1
C14H18BNO2
243.11
MFCD02179458
LIWPKYSGWSFPEE-UHFFFAOYSA-N
2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-cyanomethyl phenylboronic acid pinacol ester, 2-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, pubchem7790, 3-cyanomethylphenylboronic acid, pinacol ester, 3-cyanomethylphenyl boronic acid,pinacol ester, 3-cyanomethyl phenylboronic acid, pinacol ester
2773343
2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
CC1(C)OB(OC1(C)C)C1=CC(CC#N)=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-(Cyanomethyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 44°C to 48°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 243.11
Percent Purity 96%
Chemical Name or Material 3-(Cyanomethyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'