Få mere at vide
3-Cyanobenzyl-d6 Bromide, CDN
Artikelnummer. 30317146
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30317146 0.05 g -
30309575 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317146 Leverandør CDN Leverandørnr. CDND55680.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Cyanobenzyl Bromide-D6
CAS 1219802-21-7
Molekylær formel C8 D6 Br N
Mængde 0.05 g
Synonym 3-(Bromomethyl-d2)benzonitrile-d4, 5-(Bromomethyl-d2)benzonitrile-2,3,4,6-d4, 1-(Bromomethyl-d2)-3-cyanobenzene-2,4,5,6-d4, 3-Cyanobenzyl-d6 bromide, m-(Bromomethyl-d2)benzonitrile-d4, m-Cyano-α-bromotoluene-d6, m-Cyanobenzyl-d6 bromide, α-Bromo-m-tolunitrile-d6, 3-Cyanobenzyl Bromide-D6
InChI formel InChI=1 S/C8H6BrN/c9-5-7-2-1-3-8(4-7)6-10/h1-4 H,5H2/i1D,2 D,3 D,4 D,5D2
SMIL [2 H]c1c([2 H])c(C#N)c([2 H])c(c1[2 H])C([2 H])([2 H])Br
IUPAC navn 3-[bromo(dideuterio)methyl]-2,4,5,6-tetradeuteriobenzonitrile
Molekylvægt (g/mol) 202.0809
Formel vægt 201.006 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity