Få mere at vide
3-Cyano-5-methylhexanoic Acid, TRC
Artikelnummer. 30138621
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138621 100 mg
30006215 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138621 Leverandør Toronto Research Chemicals Leverandørnr. TRCC987565100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 181289-16-7
Molekylær formel C8H13NO2
Molekylvægt (g/mol) 155.19
Synonym Hexanoic acid, 3-cyano-5-methyl-, 3-Cyano-5-methylhexanoic acid
IUPAC navn 3-cyano-5-methylhexanoic acid
SMIL O=C(O)CC(C#N)CC(C)C
Tekniske data
Kemisk navn eller materiale 3-Cyano-5-methylhexanoic Acid
Mængde 100 mg
InChI formel InChI=1S/C8H13NO2/c1-6(2)3-7(5-9)4-8(10)11/h6-7H,3-4H2,1-2H3,(H,10,11)
Formel vægt 155.0946