missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-5-hydroxypyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-7031805
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
152803-24-2
C6H4N2O
120.11
MFCD11044307
KAXIYYPIORYZLB-UHFFFAOYSA-N
15342142
5-hydroxypyridine-3-carbonitrile
OC1=CN=CC(=C1)C#N
This item is not returnable. View return policy
152803-24-2
C6H4N2O
120.11
MFCD11044307
KAXIYYPIORYZLB-UHFFFAOYSA-N
15342142
5-hydroxypyridine-3-carbonitrile
OC1=CN=CC(=C1)C#N

3-Cyano-5-hydroxypyridine is active pharmaceutical ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Cyano-5-hydroxypyridine is active pharmaceutical ingredients.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 213°C to 217°C
Quantity 250 mg
UN Number UN3439
Synonym 5-hydroxynicotinonitrile, 3-cyano-5-hydroxypyridine, 3-pyridinecarbonitrile, 5-hydroxy, 5-hydroxy-nicotinonitrile, 5-hydroxy-3-pyridinecarbonitrile, 5-cyano-3-hydroxypyridine, pubchem17727, kaxiyypioryzlb-uhfffaoysa, 5-cyano-5-hydroxypyridine, 3-pyridinecarbonitrile,5-hydroxy
Solubility Information Slightly soluble in water.
Formula Weight 120.11
Percent Purity 97%
Chemical Name or Material 3-Cyano-5-hydroxypyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'