Learn More
Alfa Aesar™ 3-Cyano-5-fluoropyridine, 97%
Brand: Alfa Aesar™ H34292.MD
Additional Details : CAS Number : 696-42-4 Weight : 0.00025kg
Quantity | 250mg |
---|
Chemical Identifiers
696-42-4 | |
122.102 | |
LQJWCPZRNHAVJD-UHFFFAOYSA-N | |
22463471 | |
C1=C(C=NC=C1F)C#N |
C6H3FN2 | |
MFCD09027295 | |
5-fluoronicotinonitrile, 3-cyano-5-fluoropyridine, 5-fluoro-3-pyridinecarbonitrile, 3-fluoro-5-cyanopyridine, 5-fluoro-nicotinonitrile, acmc-1az9d, 3-pyridinecarbonitrile,5-fluoro, 5-fluoro-3-cyanopyridine, 5-fluoranylpyridine-3-carbonitrile | |
5-fluoropyridine-3-carbonitrile |
Specifications
3-Cyano-5-fluoropyridine | |
250mg | |
C6H3FN2 | |
5-fluoronicotinonitrile, 3-cyano-5-fluoropyridine, 5-fluoro-3-pyridinecarbonitrile, 3-fluoro-5-cyanopyridine, 5-fluoro-nicotinonitrile, acmc-1az9d, 3-pyridinecarbonitrile,5-fluoro, 5-fluoro-3-cyanopyridine, 5-fluoranylpyridine-3-carbonitrile | |
C1=C(C=NC=C1F)C#N | |
122.102 | |
122.1 |
50°C to 54°C | |
696-42-4 | |
MFCD09027295 | |
LQJWCPZRNHAVJD-UHFFFAOYSA-N | |
5-fluoropyridine-3-carbonitrile | |
22463471 | |
97% |