missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Cyano-4-iodopyridine, 95%

Brand:  Alfa Aesar™ H27603.03

Additional Details : CAS Number : 490039-72-0 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 11496520

  • 2094.00 DKK / 1g

Estimated Shipment
25-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

490039-72-0
230.008
CJKHIIQONPNAIP-UHFFFAOYSA-N
11138941
C1=CN=CC(=C1I)C#N
C6H3IN2
MFCD09037453
3-cyano-4-iodopyridine, 4-iodonicotinonitrile, 3-pyridinecarbonitrile,4-iodo, acmc-20aibr, 4-iodo-nicotinonitrile, 4-iodo-3-pyridinecarbonitrile, 4-iodanylpyridine-3-carbonitrile
4-iodopyridine-3-carbonitrile

Specifications

3-Cyano-4-iodopyridine
∽124°C (decomposition)
490039-72-0
MFCD09037453
3-cyano-4-iodopyridine, 4-iodonicotinonitrile, 3-pyridinecarbonitrile,4-iodo, acmc-20aibr, 4-iodo-nicotinonitrile, 4-iodo-3-pyridinecarbonitrile, 4-iodanylpyridine-3-carbonitrile
C1=CN=CC(=C1I)C#N
230.008
230.01
Light sensitive
1g
C6H3IN2
UN3439
CJKHIIQONPNAIP-UHFFFAOYSA-N
4-iodopyridine-3-carbonitrile
11138941
95%
Videos
SDS Documents