missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-4-fluorobenzenesulfonyl chloride, 98%, Thermo Scientific™

Catalog Number p-7047884
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
351003-23-1
C7H3ClFNO2S
219.614
MFCD03093760
KDVZADPGGPBAAK-UHFFFAOYSA-N
4-fluoro-3-cyanobenzenesulfonyl chloride, 3-cyano-4-fluorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-cyano-4-fluoro, 3-cyano-4-fluorobenzenesulphonyl chloride, 3-cyano-4-fluoro-benzenesulfonyl chloride, 4-fluoro-3-cyanobenzenesulfonylchloride, 4-fluoro-3-cyanobenzenesulphonyl chloride, 5-chlorosulfonyl-2-fluorobenzenecarbonitrile, pubchem5110, acmc-20a3ve
2778908
3-cyano-4-fluorobenzenesulfonyl chloride
C1=CC(=C(C=C1S(=O)(=O)Cl)C#N)F
This item is not returnable. View return policy
351003-23-1
C7H3ClFNO2S
219.614
MFCD03093760
KDVZADPGGPBAAK-UHFFFAOYSA-N
4-fluoro-3-cyanobenzenesulfonyl chloride, 3-cyano-4-fluorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-cyano-4-fluoro, 3-cyano-4-fluorobenzenesulphonyl chloride, 3-cyano-4-fluoro-benzenesulfonyl chloride, 4-fluoro-3-cyanobenzenesulfonylchloride, 4-fluoro-3-cyanobenzenesulphonyl chloride, 5-chlorosulfonyl-2-fluorobenzenecarbonitrile, pubchem5110, acmc-20a3ve
2778908
3-cyano-4-fluorobenzenesulfonyl chloride
C1=CC(=C(C=C1S(=O)(=O)Cl)C#N)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 67°C to 70°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 219.62
Percent Purity 98%
Chemical Name or Material 3-Cyano-4-fluorobenzenesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'