missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyano-2-pyridone, 98%, Thermo Scientific Chemicals
244.43 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store at -20°C. Keep container tightly closed. Store away from oxidizing agent.
It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store at -20°C. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Melting Point | 224°C to 226°C |
Quantity | 5 g |
UN Number | UN3439 |
Beilstein | 4661670 |
Solubility Information | Soluble in dimethyl sulfoxide. |
Formula Weight | 120.11 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 3-Cyano-2-pyridone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'