Få mere at vide
3-Cyano-2-ethyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isourea, Mikromol™
Artikelnummer. 30050482
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050482 100 mg
30051081 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050482 Leverandør Mikromol Leverandørnr. MM0020.19
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Cyano-2-ethyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isourea
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Cyano-2-ethyl-1-[2-[((5-methyl-1H-imidazol-4-yl)methyl)sulphanyl]ethyl]isourea
CAS 75850-09-8
Analyt- eller komponentnavne 3-Cyano-2-ethyl-1-[2-[((5-methyl-1H-imidazol-4-yl)methyl)sulphanyl]ethyl]isourea
Forsendelsestilstand Room Temperature
Molekylær formel C11 H17 N5 O S
Certificeringer/compliance ISO 9001
Synonym Methylethyl Analogue of Oxybutynin Hydrochloride, N-Cyano-N'-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]thio]ethyl]carbamimidic Acid Ethyl Ester (9CI), 3-Cyano-2-ethyl-1-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethyl]isourea
InChI formel InChI=1S/C11H17N5OS/c1-3-17-11(14-7-12)13-4-5-18-6-10-9(2)15-8-16-10/h8H,3-6H2,1-2H3,(H,13,14)(H,15,16)
SMIL CCOC(=NC#N)NCCSCc1nc[nH]c1C
IUPAC navn ethyl N'-cyano-N-[2-[(5-methyl-1H-imidazol-4-yl)methylsulfanyl]ethyl]carbamimidate
Molekylvægt (g/mol) 267.35
Mængde 100 mg
Formel vægt 267.351 g/mol