Få mere at vide
3-Chlorotoluene-2,4,6-d3, CDN
Artikelnummer. 30310259
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30310259 0.1 g
30320750 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310259 Leverandør CDN Leverandørnr. CDND64090.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Chlorotoluene-2,4,6-D3
CAS 1219803-79-8
Molekylær formel C7 D3 H4 Cl
Mængde 0.1 g
Synonym 3-Chlorotoluene-d3, 3-Chlorotoluene-2,4,6-d3, 3-Chlorotoluene-2,4,6-D3, 1-Chloro-3-methylbenzene-2,4,6-d3
InChI formel InChI=1 S/C7H7Cl/c1-6-3-2-4-7(8)5-6/h2-5 H,1H3/i3D,4 D,5 D
SMIL [2 H]c1cc([2 H])c(Cl)c([2 H])c1C
IUPAC navn 2-chloro-1,3,5-trideuterio-4-methylbenzene
Molekylvægt (g/mol) 129.602
Formel vægt 129.0425 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity