missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Chlorophenylacetonitrile, 99%

Brand:  Alfa Aesar™ A15673.14

Additional Details : CAS Number : 1529-41-5 Weight : 0.02500kg Transport : UN number : 3276 Chem class : 6.1 Pack group : II

Quantity 25g

 View more versions of this product

Product Code. 11497607

  • 278.00 DKK / 25g

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1529-41-5
151.593
GTIKLPYCSAMPNG-UHFFFAOYSA-N
73722
C1=CC(=CC(=C1)Cl)CC#N
C8H6ClN
MFCD00001908
3-chlorobenzyl cyanide, 3-chlorophenyl acetonitrile, 2-3-chlorophenyl acetonitrile, 3-chlorophenylacetonitrile, 3-chlorobenzeneacetonitrile, m-chlorobenzyl cyanide, acetonitrile, m-chlorophenyl, benzeneacetonitrile, 3-chloro, m-chlorophenyl acetonitrile, 2-3-chlorophenyl ethanenitrile
2-(3-chlorophenyl)acetonitrile

Specifications

3-Chlorophenylacetonitrile
>110°C (230°F)
1.5437
11°C to 13°C
1529-41-5
MFCD00001908
2042965
GTIKLPYCSAMPNG-UHFFFAOYSA-N
2-(3-chlorophenyl)acetonitrile
73722
99%
1.189
Pungent
134°C to 136°C (10mmHg)
25g
C8H6ClN
UN3276
3-chlorobenzyl cyanide, 3-chlorophenyl acetonitrile, 2-3-chlorophenyl acetonitrile, 3-chlorophenylacetonitrile, 3-chlorobenzeneacetonitrile, m-chlorobenzyl cyanide, acetonitrile, m-chlorophenyl, benzeneacetonitrile, 3-chloro, m-chlorophenyl acetonitrile, 2-3-chlorophenyl ethanenitrile
C1=CC(=CC(=C1)Cl)CC#N
151.593
151.6
Videos
SDS Documents