Få mere at vide
3-chlorooxolane, TRC
Artikelnummer. 30353493
Skift visning
Mængde:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30353493 25 mg
30359385 250 mg
30359568 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30353493 Leverandør Toronto Research Chemicals Leverandørnr. TRCC61103025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 19311-38-7
Molekylær formel C4H7OCl
Molekylvægt (g/mol) 106.55
IUPAC navn 3-chlorooxolane
SMIL ClC1CCOC1
Tekniske data
Kemisk navn eller materiale 3-chlorooxolane
Mængde 25 mg
InChI formel InChI=1S/C4H7ClO/c5-4-1-2-6-3-4/h4H,1-3H2
Formel vægt 106.0185