missing translation for 'onlineSavingsMsg'
Learn More

3-(chloromethyl)-5-(3-thienyl)-1,2,4-oxadiazole, Thermo Scientific™

Catalog Number p-4391259
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
184970-24-9
C7H5ClN2OS
200.64
KHJYOJVUQYWASE-UHFFFAOYSA-N
3-chloromethyl-5-thiophen-3-yl-1,2,4-oxadiazole, 3-chloromethyl-5-3-thienyl-1,2,4-oxadiazole, 3-chloromethyl-5-thien-3-yl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-3-thienyl, 3-3-chloromethyl-1,2,4-oxadiazol-5-yl thiophene
2797043
3-(chloromethyl)-5-thiophen-3-yl-1,2,4-oxadiazole
C1=CSC=C1C2=NC(=NO2)CCl
This item is not returnable. View return policy
184970-24-9
C7H5ClN2OS
200.64
KHJYOJVUQYWASE-UHFFFAOYSA-N
3-chloromethyl-5-thiophen-3-yl-1,2,4-oxadiazole, 3-chloromethyl-5-3-thienyl-1,2,4-oxadiazole, 3-chloromethyl-5-thien-3-yl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-3-thienyl, 3-3-chloromethyl-1,2,4-oxadiazol-5-yl thiophene
2797043
3-(chloromethyl)-5-thiophen-3-yl-1,2,4-oxadiazole
C1=CSC=C1C2=NC(=NO2)CCl
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 200.65
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)-5-(3-thienyl)-1, 2, 4-oxadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'