missing translation for 'onlineSavingsMsg'
Learn More

3-Chloromethyl-5-(3-methoxyphenyl)-1,2,4-oxadiazole, 99%, Thermo Scientific™

Catalog Number p-7033188
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
175205-62-6
C10H9ClN2O2
224.64
MFCD00084966
GSWMZQIUADKXLN-UHFFFAOYSA-N
3-chloromethyl-5-3-methoxyphenyl-1,2,4-oxadiazole, acmc-1bxk1, 3-chloromethyl-5-3-methoxyphenyl-1,2-4-oxadiazole, 3-chloromethyl-5-3-methoxy-phenyl-1,2,4 oxadiazole, 3-chloromethyl-5-3-methoxyphenyl-1,2,4 oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-3-methoxyphenyl, 1-3-chloromethyl 1,2,4-oxadiazol-5-yl-3-methoxybenzene
11715562
3-(chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole
COC1=CC=CC(=C1)C1=NC(CCl)=NO1
This item is not returnable. View return policy
175205-62-6
C10H9ClN2O2
224.64
MFCD00084966
GSWMZQIUADKXLN-UHFFFAOYSA-N
3-chloromethyl-5-3-methoxyphenyl-1,2,4-oxadiazole, acmc-1bxk1, 3-chloromethyl-5-3-methoxyphenyl-1,2-4-oxadiazole, 3-chloromethyl-5-3-methoxy-phenyl-1,2,4 oxadiazole, 3-chloromethyl-5-3-methoxyphenyl-1,2,4 oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-3-methoxyphenyl, 1-3-chloromethyl 1,2,4-oxadiazol-5-yl-3-methoxybenzene
11715562
3-(chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole
COC1=CC=CC(=C1)C1=NC(CCl)=NO1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 58°C to 59°C
Quantity 250 mg
Formula Weight 224.65
Percent Purity 99%
Chemical Name or Material 3-Chloromethyl-5-(3-methoxyphenyl)-1,2,4-oxadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'