missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-5-(2-thienyl)-1,2,4-oxadiazole, ≥95%, Thermo Scientific™

Catalog Number p-2699183
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
306936-06-1
C7H5ClN2OS
200.64
MFCD01571188
YVYZVJRSESVBCM-UHFFFAOYSA-N
3-chloromethyl-5-thiophen-2-yl-1,2,4-oxadiazole, 3-chloromethyl-5-thien-2-yl-1,2,4-oxadiazole, 3-chloromethyl-5-2-thienyl-1,2,4-oxadiazole, 3-chloromethyl-5-thiophen-2-yl-1,2,4 oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-2-thienyl, 2-3-chloromethyl-1,2,4-oxadiazol-5-yl thiophene
2798343
3-(chloromethyl)-5-thiophen-2-yl-1,2,4-oxadiazole
C1=CSC(=C1)C2=NC(=NO2)CCl
This item is not returnable. View return policy
306936-06-1
C7H5ClN2OS
200.64
MFCD01571188
YVYZVJRSESVBCM-UHFFFAOYSA-N
3-chloromethyl-5-thiophen-2-yl-1,2,4-oxadiazole, 3-chloromethyl-5-thien-2-yl-1,2,4-oxadiazole, 3-chloromethyl-5-2-thienyl-1,2,4-oxadiazole, 3-chloromethyl-5-thiophen-2-yl-1,2,4 oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-2-thienyl, 2-3-chloromethyl-1,2,4-oxadiazol-5-yl thiophene
2798343
3-(chloromethyl)-5-thiophen-2-yl-1,2,4-oxadiazole
C1=CSC(=C1)C2=NC(=NO2)CCl
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 200.65
Percent Purity 95%
Chemical Name or Material 3-(Chloromethyl)-5-(2-thienyl)-1, 2, 4-oxadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'