Få mere at vide
3-Chlorocatechol, TRC
Artikelnummer. 30352617
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30352617 5 mg
30354840 10 mg
30367317 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30352617 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3735755MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4018-65-9
Molekylær formel C6H5O2Cl
Molekylvægt (g/mol) 144.55
IUPAC navn 3-chlorobenzene-1,2-diol
SMIL Oc1cccc(Cl)c1O
Tekniske data
Kemisk navn eller materiale 3-Chlorocatechol
Mængde 5 mg
InChI formel InChI=1S/C6H5ClO2/c7-4-2-1-3-5(8)6(4)9/h1-3,8-9H
Formel vægt 143.9978