missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Chlorobenzoyl chloride, 97%

Brand:  Alfa Aesar™ A13509.36

Additional Details : CAS Number : 618-46-2 Weight : 0.50000kg Transport : UN number : 3265 Chem class : 8 Pack group : II

Quantity 500g

 View more versions of this product

Product Code. 11495603

  • 2707.00 DKK / 500g

Estimated Shipment
20-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

618-46-2
175.008
WHIHIKVIWVIIER-UHFFFAOYSA-N
69252
C1=CC(=CC(=C1)Cl)C(=O)Cl
C7H4Cl2O
MFCD00000671
m-chlorobenzoyl chloride, benzoyl chloride, 3-chloro, benzoyl chloride, m-chloro, m-chlorobenzoylchloride, 3-chlorobenzoylchloride, benzoylchloride, 3-chloro, mcoc, 3-chlorobenzoic acid chloride, ccris 8615, 3chlorobenzoyl chloride
3-chlorobenzoyl chloride

Specifications

3-Chlorobenzoyl chloride
117°C (242°F)
1.569
224°C to 225°C
618-46-2
MFCD00000671
386256
WHIHIKVIWVIIER-UHFFFAOYSA-N
3-chlorobenzoyl chloride
69252
97%
1.368
Acidic
Moisture sensitive
500g
C7H4Cl2O
UN3265
m-chlorobenzoyl chloride, benzoyl chloride, 3-chloro, benzoyl chloride, m-chloro, m-chlorobenzoylchloride, 3-chlorobenzoylchloride, benzoylchloride, 3-chloro, mcoc, 3-chlorobenzoic acid chloride, ccris 8615, 3chlorobenzoyl chloride
C1=CC(=CC(=C1)Cl)C(=O)Cl
175.008
175.01
Videos
SDS Documents