Få mere at vide
3-Chloro Fenofibric Acid-d6, TRC
Artikelnummer. 30352446
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30352446 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30352446 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36468210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Chloro Fenofibric Acid-d6
CAS 1346603-07-3
Molekylær formel C17H9D6ClO4
Mængde 10 mg
Synonym 2-[4-(3-Chlorobenzoyl)phenoxy]-2-methylpropanoic Acid-d6, LF 406-d6
InChI formel InChI=1S/C17H15ClO4/c1-17(2,16(20)21)22-14-8-6-11(7-9-14)15(19)12-4-3-5-13(18)10-12/h3-10H,1-2H3,(H,20,21)/i1D3,2D3
SMIL [2H]C([2H])([2H])C(Oc1ccc(cc1)C(=O)c2cccc(Cl)c2)(C(=O)O)C([2H])([2H])[2H]
IUPAC navn 2-[4-(3-chlorobenzoyl)phenoxy]-3,3,3-trideuterio-2-(trideuteriomethyl)propanoic acid
Molekylvægt (g/mol) 324.79
Formel vægt 324.1035