missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-alpha-toluenesulfonyl chloride, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3-Chloro-alpha-toluenesulfonyl chloride is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chloro-alpha-toluenesulfonyl chloride is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents. It is sensitive to moisture.
3-Chloro-alpha-toluenesulfonyl chloride is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents. It is sensitive to moisture.
Specifications
Melting Point | 69°C to 75°C |
Quantity | 1 g |
UN Number | UN3261 |
Formula Weight | 225.09 |
Percent Purity | 96% |
Chemical Name or Material | 3-Chloro-alpha-toluenesulfonyl chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'