missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-chloro-5-fluorobenzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
20 g
Packaging:
Glass bottle
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Linear Formula | FC6H3(Cl)CHO |
Quantity | 5 g |
Formula Weight | 158.56 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'