missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(tert-butylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific™

Catalog Number p-7035352
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
850589-46-7
C11H15BClNO3
255.505
MFCD07363771
RFTCLOPBDGFPDR-UHFFFAOYSA-N
4-tert-butylcarbamoyl-3-chlorophenyl boronic acid, 3-chloro-4-n-tert-butylcarbamoyl phenylboronic acid, 4-tert-butylcarbamoyl-3-chlorophenylboronic acid, 3-chloro-4-n-tert-butylcarbamoyl benzeneboronic acid, 3-chloro-4-tert-butylcarbamoyl benzeneboronic acid, n-tert-butyl 2-chloro-4-boronobenzamide, n-tert-butyl 4-borono-2-chlorobenzamide, 4-tert-butylcarbamoyl-3-chlorophenyl boronicacid, boronic acid,b-3-chloro-4-1,1-dimethylethyl amino carbonyl phenyl
44119651
[4-(tert-butylcarbamoyl)-3-chlorophenyl]boronic acid
B(C1=CC(=C(C=C1)C(=O)NC(C)(C)C)Cl)(O)O
This item is not returnable. View return policy
850589-46-7
C11H15BClNO3
255.505
MFCD07363771
RFTCLOPBDGFPDR-UHFFFAOYSA-N
4-tert-butylcarbamoyl-3-chlorophenyl boronic acid, 3-chloro-4-n-tert-butylcarbamoyl phenylboronic acid, 4-tert-butylcarbamoyl-3-chlorophenylboronic acid, 3-chloro-4-n-tert-butylcarbamoyl benzeneboronic acid, 3-chloro-4-tert-butylcarbamoyl benzeneboronic acid, n-tert-butyl 2-chloro-4-boronobenzamide, n-tert-butyl 4-borono-2-chlorobenzamide, 4-tert-butylcarbamoyl-3-chlorophenyl boronicacid, boronic acid,b-3-chloro-4-1,1-dimethylethyl amino carbonyl phenyl
44119651
[4-(tert-butylcarbamoyl)-3-chlorophenyl]boronic acid
B(C1=CC(=C(C=C1)C(=O)NC(C)(C)C)Cl)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 130°C to 134°C
Quantity 250 mg
Formula Weight 255.51
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-(tert-butylcarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'