Få mere at vide
3-Chloro-2-methylbiphenyl-d5, TRC
Artikelnummer. 30354362
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30354362 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30354362 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36910710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Chloro-2-methylbiphenyl-d5
CAS 1330173-04-0
Molekylær formel C13H6D5Cl
Mængde 10 mg
Synonym 3-Chloro-2-methyl-1,1'-biphenyl-d5
InChI formel InChI=1S/C13H11Cl/c1-10-12(8-5-9-13(10)14)11-6-3-2-4-7-11/h2-9H,1H3/i2D,3D,4D,6D,7D
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2cccc(Cl)c2C
IUPAC navn 1-(3-chloro-2-methylphenyl)-2,3,4,5,6-pentadeuteriobenzene
Molekylvægt (g/mol) 207.71
Formel vægt 207.0863