missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluorobenzoic acid, 97%, Thermo Scientific™

Catalog Number p-7025225
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy
161957-55-7
C7H4ClFO2
174.555
MFCD00042506
FCSSYEWURMTUSM-UHFFFAOYSA-N
2-fluoro-3-chlorobenzoic acid, 3-chloro-2-fluoro benzoic acid, 3-chloro-2-fluoro-benzoic acid, 3-chloro-2-fluorobenzoicacid, 3-chloro-2-fluorbenzoic acid, pubchem1332, acmc-209dnj, intermediates-zcf02219, benzoic acid, chlorofluoro, ksc112c3l
302931
3-chloro-2-fluorobenzoic acid
C1=CC(=C(C(=C1)Cl)F)C(=O)O
This item is not returnable. View return policy
161957-55-7
C7H4ClFO2
174.555
MFCD00042506
FCSSYEWURMTUSM-UHFFFAOYSA-N
2-fluoro-3-chlorobenzoic acid, 3-chloro-2-fluoro benzoic acid, 3-chloro-2-fluoro-benzoic acid, 3-chloro-2-fluorobenzoicacid, 3-chloro-2-fluorbenzoic acid, pubchem1332, acmc-209dnj, intermediates-zcf02219, benzoic acid, chlorofluoro, ksc112c3l
302931
3-chloro-2-fluorobenzoic acid
C1=CC(=C(C(=C1)Cl)F)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 176°C to 178°C
Quantity 0.25 g
Beilstein 7127637
Formula Weight 174.56
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluorobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'