missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15487516
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1192025-01-6
C12H15BClFO2
256.51
MFCD12405368
NTHMFEYDRUMMGR-UHFFFAOYSA-N
2-3-chloro-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-chloro-2-fluorophenylboronic acid pinacol ester, 3-chloro-2-fluorophenyl boronic acid pinacol ester, 3-chloro-2-fluorobenzeneboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-3-chloro-2-fluorophenyl-4,4,5,5-tetramethyl
56973159
2-(3-chloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=C(F)C(Cl)=CC=C1
This item is not returnable. View return policy
1192025-01-6
C12H15BClFO2
256.51
MFCD12405368
NTHMFEYDRUMMGR-UHFFFAOYSA-N
2-3-chloro-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-chloro-2-fluorophenylboronic acid pinacol ester, 3-chloro-2-fluorophenyl boronic acid pinacol ester, 3-chloro-2-fluorobenzeneboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-3-chloro-2-fluorophenyl-4,4,5,5-tetramethyl
56973159
2-(3-chloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=C(F)C(Cl)=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 256.51
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Chloro-2-fluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'