missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloro-2'-fluoroacetophenone, 96%, Thermo Scientific™

Catalog Number p-7029042
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
161957-59-1
C8H6ClFO
172.583
MFCD00106165
ZHDLNEIXOUIAQO-UHFFFAOYSA-N
1-3-chloro-2-fluorophenyl ethanone, 3'-chloro-2'-fluoroacetophenone, 1-acetyl-3-chloro-2-fluorobenzene, ethanone, 1-chlorofluorophenyl, 1-3-chloro-2-fluorophenyl ethan-1-one, 1-3-chloro-2-fluoro-phenyl ethanone, acmc-1ch2t, 3-chloro-2-fluoroacetophenone, 2'-fluoro-3'-chloroacetophenone, ethanone,1-3-chloro-2-fluorophenyl
11116531
1-(3-chloro-2-fluorophenyl)ethanone
CC(=O)C1=C(C(=CC=C1)Cl)F
This item is not returnable. View return policy
161957-59-1
C8H6ClFO
172.583
MFCD00106165
ZHDLNEIXOUIAQO-UHFFFAOYSA-N
1-3-chloro-2-fluorophenyl ethanone, 3'-chloro-2'-fluoroacetophenone, 1-acetyl-3-chloro-2-fluorobenzene, ethanone, 1-chlorofluorophenyl, 1-3-chloro-2-fluorophenyl ethan-1-one, 1-3-chloro-2-fluoro-phenyl ethanone, acmc-1ch2t, 3-chloro-2-fluoroacetophenone, 2'-fluoro-3'-chloroacetophenone, ethanone,1-3-chloro-2-fluorophenyl
11116531
1-(3-chloro-2-fluorophenyl)ethanone
CC(=O)C1=C(C(=CC=C1)Cl)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.5315
Quantity 1 g
Formula Weight 172.59
Percent Purity 96%
Chemical Name or Material 3'-Chloro-2'-fluoroacetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'