missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluoro-6-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Catalog Number 15438225
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1092461-38-5
C7H4ClF4N
213.56
MFCD11519381
ZHDSWEWAMCTWAZ-UHFFFAOYSA-N
3-chloro-2-fluoro-6-trifluoromethyl aniline, 2-amino-4-chloro-3-fluorobenzotrifluoride, 3-chloro-2-fluoro-6-trifluoromethyl benzenamine, 3-chloro-2-fluoro-6-trifluoromethyl phenylamine, 3-chloro-2-fluoro-6-trifluoromethyl aniline, jrd
57361729
3-chloro-2-fluoro-6-(trifluoromethyl)aniline
C1=CC(=C(C(=C1C(F)(F)F)N)F)Cl
This item is not returnable. View return policy
1092461-38-5
C7H4ClF4N
213.56
MFCD11519381
ZHDSWEWAMCTWAZ-UHFFFAOYSA-N
3-chloro-2-fluoro-6-trifluoromethyl aniline, 2-amino-4-chloro-3-fluorobenzotrifluoride, 3-chloro-2-fluoro-6-trifluoromethyl benzenamine, 3-chloro-2-fluoro-6-trifluoromethyl phenylamine, 3-chloro-2-fluoro-6-trifluoromethyl aniline, jrd
57361729
3-chloro-2-fluoro-6-(trifluoromethyl)aniline
C1=CC(=C(C(=C1C(F)(F)F)N)F)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.4865
Quantity 250 mg
UN Number UN2810
Formula Weight 213.56
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluoro-6-(trifluoromethyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'